Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde ≥98%
Benzaldehyde ≥98%
Benzaldehyde ≥98%
Benzaldehyde ≥98%
Purity: 98% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299
  •  :100-52-7
  •  :C₆H₅CHO
  •  :MFCD00003299
  •  :106.12 g/mol
  •  :14,1058
  •  :178…179 °C
  •  :–26 °C
  •  :1.047
  •  :62 °C (143 °F)
  •  :Ambient
  •  :202-860-4
  •  :1990
  •  :9,III
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    250 g
    1 * 250 g
    1 * 250 g - 196 zł
    196 zł
    1 * 250 g
       196 zł
  • Glass bottle for solids
    1 kg
    1 * 1 kg
    1 * 1 kg - 274 zł
    274 zł
    1 * 1 kg
       274 zł
  • 3 kg
    1 * 3 kg
    1 * 3 kg - 658 zł
    658 zł
    1 * 3 kg
       658 zł