Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitrophenylhydrazine
2-Nitrophenylhydrazine, Sigma-Aldrich®
2-Nitrophenylhydrazine, Sigma-Aldrich®
SIALN21588-10G
:
Merck
:
2-Nitrophenylhydrazine, Sigma-Aldrich®
SIALN21588-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
153.14 g/mol
:
4.1,II
:
M-Clarity™ quality level = MQ100
:
221-222-6
:
2926
:
3034-19-3
:
C₆H₇N₃O₂