2-Hydroxy-4-(trifluoromethyl)pyridine, Sigma-Aldrich®
Catalog # SIAL648876-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:163.1 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:50650-59-4
Formula:C₆H₄F₃NO
MDL Number:MFCD00153195