Order Entry
Poland
ContactUsLinkComponent
2-Ethylphenylhydrazine hydrochloride, Sigma-Aldrich®
2-Ethylphenylhydrazine hydrochloride, Sigma-Aldrich®
Catalog # SIAL393916-10G
Supplier:  Merck
CAS Number:  
2-Ethylphenylhydrazine hydrochloride, Sigma-Aldrich®
Catalog # SIAL393916-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :172.66 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:421-460-2
CAS Number:19398-06-2
 :58711-02-7
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00071599