2,6-Toluenediisocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:91-08-7
:
Formula:C₉H₆N₂O₂
MDL Number:MFCD00002010
:174.16
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:202-039-0
UN:2078
Frequently Bought Together