

2,4-Toluenodiizocyjanian ≥80%, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:584-84-9
:26471-62-5
Formula:C₉H₆N₂O₂
MDL Number:MFCD00002011
:174.16
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:247-722-4
UN:2078
:GHS06
:GHS08
Storage Temperature:Temperatura otoczenia
Specification Test Results
- Appearance (Color):Colorless or White
- Appearance (Form):Conforms to Requirements (Liquid, Crystals or Solid)
- Infrared spectrum:Conforms to Structure
- Tolylene-2,4-diisocyanate (GC):> 78.0 %
- Product of Supplier:Confirmed (BASF AG (Aktiengesellschaft)
Specification Test Results
Frequently Bought Together