
Sulpholane ≥99.0% for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:126-33-0
:
Formula:C₄H₈O₂S
MDL Number:MFCD00005484
:120.17 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:26 °C
Density:1.26 g/cm3 (30 °C)
Flash Pt:177 °C
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:204-783-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC, area%):≥ 99.0 % (a/a)
- Water (K. F.):≤ 0.20 %
- Identity (IR):passes test
- Boiling Point:285 °C (1013 hPa)
- Vapor Pressure:0.035 hPa (30 °C)
- Flash Point:177 °C
- Refractive Index:1.484 (20 °C, 589 nm)
- Density:1.26 g/cm3 (30 °C)
Specification Test Results
Frequently Bought Together