
(S)-(-)-3-Amino-1,2-propanediol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61278-21-5
:
Formula:C₃H₉NO₂
MDL Number:MFCD00798260
:91.11 g/mol
Boiling Pt:117…119 °C (0.4 mmHg)
Melting Pt:54…56 °C
Density:1.175
Flash Pt:>110 °C (230 °F)
ADR:8,III
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together