
Ammonium hexachlororuthenate(IV) (≥28.4% Ru)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18746-63-9
:
Formula:H₈Cl₆N₂Ru
MDL Number:MFCD00015962
:349.86 g/mol
Storage Temperature:Ambient
EINECS:242-552-7
UN:3288
ADR:6.1,III
:GHS06
Related Information
Frequently Bought Together