Thiocarbonyl chloride 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:463-71-8
:
Formula:CCl₂S
MDL Number:MFCD00004918
:114.98 g/mol
Boiling Pt:73 °C (760 mmHg)
Density:1.508 g/cm³
Flash Pt:62 °C
Storage Temperature:Refrigerator
EINECS:207-341-6
UN:2474
ADR:6.1,
:GHS06
Specification Test Results
- Appearance:Orange-red to red Liquid
- GC:≥80 %
- Melting point:164 to 169 °C
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together