Order Entry
Poland
ContactUsLinkComponent
1-Bromo-2,4-difluorobenzene 98+%
Irritant
Flammable
1-Bromo-2,4-difluorobenzene 98+%
Certificates
undefined
Irritant
Flammable
1-Bromo-2,4-difluorobenzene 98+%

Specification Test Results

  • Appearance:
    Clear colorless to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5040 to 1.5060 (20°C, 589 nm)
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together