
3-Bromobenzoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:585-76-2
:
Formula:C₇H₅BrO₂
MDL Number:MFCD00002487
:201.02 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:154…158 °C
Density:1,845 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-562-3
:GHS07
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together