Order Entry
Poland
ContactUsLinkComponent
 
1,4-Dibromobenzene
Catalog # MOLEM33944638
Supplier:  Molekula
CAS Number:  
1,4-Dibromobenzene
Catalog # MOLEM33944638
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:13,03042
Boiling Pt:219 °C (1013 hPa)
Melting Pt:84…87 °C
Density:2.28 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:203-390-2
UN:3077
ADR:9,III
 :235.91 g/mol
CAS Number:106-37-6
Formula:C₆H₄Br₂
MDL Number:MFCD00000089