
1,4-Dibromobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-37-6
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000089
Merck Index:14,3019
Boiling Pt:219…220 °C
Melting Pt:86…90 °C
Density:1.841
Flash Pt:>100 °C (212 °F)
UN:3077
ADR:9,III
:235.91 g/mol
EINECS:203-390-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together