
1,3-Dibromobenzene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-36-1
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000078
:235.91 g/mol
Boiling Pt:218…219 °C (760 mmHg)
Melting Pt:–7 °C
Density:1.95 g/cm³
Flash Pt:93 °C
EINECS:203-574-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6070 to 1.6090 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together