Order Entry
Poland
ContactUsLinkComponent
1,2-Dibromobenzene
1,2-Dibromobenzene
Catalog # MOLEM50788561
Supplier:  Molekula
CAS Number:  
1,2-Dibromobenzene
Catalog # MOLEM50788561
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :235.91 g/mol
Boiling Pt:224 °C (1013 hPa)
Density:1.956 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:209-507-3
UN:3082
ADR:9,III
CAS Number:583-53-9
Formula:C₆H₄Br₂
MDL Number:MFCD00000057