1-Octene 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-66-0
:
Formula:C₈H₁₆
MDL Number:MFCD00009548
Boiling Pt:121…123 °C (760 mmHg)
Melting Pt:–102 °C
Density:0.715 g/cm³
Molecular Weight:112.22 g/mol
Flash Pt:13 °C
Storage Temperature:Ambient
UN:3295
ADR:3 II
Merck Index:13,01770
EINECS:203-893-7
:GHS02
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4080 to 1.4100 (20°C, 589 nm)
- Water:≤0.005 %
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together