




cis-1,4-Dichlorobut-2-en 95%
Catalog # ACRO187600250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Rodzaj opakowania:Glass bottle
- Ilość:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4884 to 1.4914 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
About this item
:
Boiling Pt:152 °C (758 mmHg)
Melting Pt:–48 °C
Density:1,18 g/cm³
Flash Pt:55 °C
Storage Temperature:Lodówka
EINECS:216-021-5
UN:3489
ADR:6.1,
:125 g/mol
:GHS06
:GHS08
:GHS05
:GHS09
:GHS02
CAS Number:1476-11-5
Formula:C₄H₆Cl₂
MDL Number:MFCD00062950