Squalane ≥99.0% (by GC) for gas chromatography, Fluka™
85629-50ML
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Identity (IR):complying
- Identity (NMR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.451 - 1.453
:
:422.82 g/mol
:12,08923
:176 °C (0.07 hPa)
:-38 °C
:–38 °C
:0.82 g/cm³ (20 °C)
:218 °C
:Ambient
:203-825-6
:111-01-3
:C₃₀H₆₂
:MFCD00008953