Złóż zamówienie
Poland
ContactUsLinkComponent
tert-butyloamina do syntezy, Sigma-Aldrich®
tert-butyloamina do syntezy, Sigma-Aldrich®
Supplier:  Merck
Certificates
tert-butyloamina do syntezy, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -72 °C
  • Molar Mass:
    73.14 g/mol
  • Boiling Point:
    44 °C
  • Vapor Pressure:
    300 hPa (20 °C)
  • Flash Point:
    -38 °C DIN 51755 Part 1
  • Refractive Index:
    1.37 (20 °C, 589 nm)
  • Explosion Limit:
    1.5 - 9.2 %(V)
  • Density:
    0.69 g/cm3 (20 °C)
  • pH:
    12 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    375 °C DIN 51794

Specification Test Results

Related Information

Frequently Bought Together