

1-Bromo-2-methylpropane 98+%
:78-77-3
:
:C₄H₉Br
:MFCD00000217
:137.02 g/mol
:13,05152
:90…92 °C (760 mmHg)
:–119 °C
:1.26 g/cm³
:18 °C
:Ambient
:201-141-2
:2342
:3,II
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4370 (20°C, 589 nm)
- GC:≥98.0 %
Frequently Bought Together