1,2-Dibromo-2-methylpropane 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:594-34-3
- Formula:C₄H₈Br₂
- MDL Number:MFCD00000126
- Molecular Weight:215.92 g/mol
- Boiling Pt:150 °C (760 mmHg)
- Melting Pt:-70,1 °C
- Density:1.79 g/cm³
- Flash Pt:>110 °C
- :Density: 1.7675 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:209-835-7
3 Options Available Below
Product Details & Documents
- CAS Number:594-34-3
- Formula:C₄H₈Br₂
- MDL Number:MFCD00000126
- Molecular Weight:215.92 g/mol
- Boiling Pt:150 °C (760 mmHg)
- Melting Pt:-70,1 °C
- Density:1.79 g/cm³
- Flash Pt:>110 °C
- :Density: 1.7675 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:209-835-7
Documents
ACRO40649
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5083 to 1.5103 (20°C, 589 nm)
- GC:≥97.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



