Syringe filter holder, stainless steel
Catalog # SART16214
Supplier: Sartorius
Specifications
- For Ø:25 mm
- Material:Stainless steel
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This syringe filter holder is made from stainless steel. PTFE coated surface on the top provides leak proof sealing, and the chemical compatibility depends upon the filter inserted.
- Re-usable
- Resistant to heat