Order Entry
Poland
ContactUsLinkComponent
n-Octane
n-Octane
Catalog # PROOCHE124
CAS Number:  
undefined
n-Octane
Catalog # PROOCHE124
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :114.23 g/mol
Merck Index:13,06782
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–57 °C
Density:0.703 g/cm³ (20 °C)
Flash Pt:8 °C
Storage Temperature:Ambient
EINECS:203-892-1
UN:1262
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:111-65-9
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556