Order Entry
Poland
ContactUsLinkComponent
DL-α-Tocopherol (Vitamin E), Sigma-Aldrich®
DL-α-Tocopherol (Vitamin E), Sigma-Aldrich®
Catalog # SIAL90669-50G-F
Supplier:  Merck
CAS Number:  
DL-α-Tocopherol (Vitamin E), Sigma-Aldrich®
Catalog # SIAL90669-50G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :430.71 g/mol
EINECS:233-466-0
CAS Number:10191-41-0
 :1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051