Order Entry
Poland
ContactUsLinkComponent
Phenyl propargyl ether, Sigma-Aldrich®
Phenyl propargyl ether, Sigma-Aldrich®
Catalog # SIAL78960-10ML
Supplier:  Merck
CAS Number:  
Phenyl propargyl ether, Sigma-Aldrich®
Catalog # SIAL78960-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :132.16 g/mol
EINECS:237-095-5
CAS Number:13610-02-1
Formula:C₉H₈O
MDL Number:MFCD00048107