Propagyl bromide ∼9.2 mol/l in xylene ≥80%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-96-7
:
Formula:C₃H₃Br
MDL Number:MFCD00000241
:118.96 g/mol
:M-Clarity™ quality level = MQ100
EINECS:203-447-1
Frequently Bought Together