1H,1H-Nonadecafluoro-1-decanol, Sigma-Aldrich®
Catalog # SIAL446831-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:500.1 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:307-37-9
Formula:C₁₀H₃F₁₉O
MDL Number:MFCD00153234