1-Decyl-2-methylimidazole, Sigma-Aldrich®
Catalog # SIAL433799-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:222.37 g/mol
:M-Clarity™ quality level = MQ100
EINECS:255-633-7
CAS Number:42032-30-4
Formula:C₁₄H₂₆N₂