Order Entry
Poland
ContactUsLinkComponent
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, Sigma-Aldrich®
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, Sigma-Aldrich®
Catalog # SIAL406422-5G
Supplier:  Merck
CAS Number:  
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, Sigma-Aldrich®
Catalog # SIAL406422-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :262.1 g/mol
Boiling Pt:135…136 °C (27 mmHg)
Melting Pt:69 °C
 :M-Clarity™ quality level = MQ100
CAS Number:355-74-8
Formula:C₆H₆F₈O₂
MDL Number:MFCD00069087