1H,1H,2H,2H-Tridecafluoro-1-octanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:647-42-7
:
Formula:C₈H₅F₁₃O
MDL Number:MFCD00042143
:364.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:211-477-1
Frequently Bought Together