1H,1H-Pentadecafluoro-1-octanol, Sigma-Aldrich®
Catalog # SIAL257451-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:400.09 g/mol
:M-Clarity™ quality level = MQ200
EINECS:206-197-1
CAS Number:307-30-2
Formula:C₈H₃F₁₅O
MDL Number:MFCD00004675