1H,1H-Tricosafluoro-1-dodecanol tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:423-65-4
:
Formula:C₁₂H₃F₂₃O
MDL Number:MFCD00153235
:600.12 g/mol
Boiling Pt:224 °C (740 mmHg)
Melting Pt:∼ 110 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together