
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:647-42-7
:
Formula:C₈H₅F₁₃O
MDL Number:MFCD00042143
:364.11 g/mol
Boiling Pt:90 °C (20 mmHg)
Density:1.68
Flash Pt:91 °C (195 °F)
EINECS:211-477-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together