Order Entry
Poland
ContactUsLinkComponent
1H,1H,7H-Dodecafluoro-1-heptanol ≥97%
1H,1H,7H-Dodecafluoro-1-heptanol ≥97%
Catalog # B20144.14
CAS Number:  
1H,1H,7H-Dodecafluoro-1-heptanol ≥97%
Catalog # B20144.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

CAS Number:335-99-9
 :
Formula:C₇H₄F₁₂O
MDL Number:MFCD00039630
 :332.09 g/mol
Boiling Pt:169…170 °C
Melting Pt:–20 °C
Density:1.762
Storage Temperature:Ambient
EINECS:206-406-6