

Phenyl propargyl ether ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13610-02-1
:
Formula:C₉H₈O
MDL Number:MFCD00048107
:132.16 g/mol
Boiling Pt:89…90 °C (14 mmHg)
Density:1.030
Flash Pt:76 °C (169 °F)
EINECS:237-095-5
:GHS05
:GHS07
Related Information
Frequently Bought Together