

Phenyl propargyl ether 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13610-02-1
:
Formula:C₉H₈O
MDL Number:MFCD00048107
:132.16 g/mol
Boiling Pt:89…90 °C (14 mmHg)
Density:1.03 g/cm³
Flash Pt:76 °C
EINECS:237-095-5
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5320 to 1.5350 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
Frequently Bought Together