Order Entry
Poland
ContactUsLinkComponent
1H,1H-Pentadecafluoro-1-octanol 98%
Irritant
1H,1H-Pentadecafluoro-1-octanol 98%
Certificates
undefined
Irritant
1H,1H-Pentadecafluoro-1-octanol 98%

Specification Test Results

  • Appearance:
    White Crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    43 to 47 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together