n-Heptadecane, Supelco®
Catalog # 51578-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:240.47 g/mol
Boiling Pt:302.1 °C (1013 hPa)
Melting Pt:21.2 °C
Density:0.7783 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:148 °C
Storage Temperature:Ambient
EINECS:211-108-4
:GHS08
CAS Number:629-78-7
Formula:H₃C(CH₂)₁₅CH₃
MDL Number:MFCD00009002