n-Heptadecane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-78-7
:
Formula:H₃C(CH₂)₁₅CH₃
MDL Number:MFCD00009002
:240.47 g/mol
Boiling Pt:302.1 °C (1013 hPa)
Melting Pt:21.2 °C
Density:0.7783 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:148 °C
EINECS:211-108-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:22 °C
- Molar Mass:240.47 g/mol
- Boiling Point:302 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:148 °C
- Density:0.78 g/cm3 (20 °C)
- Viscosity Kinematic:- 7 mm2/s (40 °C)
Specification Test Results
Frequently Bought Together