Sodium dodecyl sulphate (SDS) 98% for biochemistry, for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Molecular Weight:288.38 g/mol
Melting Pt:206 °C
Flash Pt:>150 °C
ADR:4.1 III
Merck Index:14,08636
EINECS:205-788-1
UN:1325
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or flakes
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (1 g, 105°C, 3 h)
- Solubility:(3 % in water) Clear colorless
- GC:≥97.5 %
- UV:(3 % in water)
- Chloride (Cl):≤500 ppm
Specification Test Results
Frequently Bought Together