
Sodium decyl sulphate for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-87-0
:
Formula:C₁₀H₂₁NaO₄S
MDL Number:MFCD00041881
:260.33 g/mol
Melting Pt:191,85 °C
Density:1,207 g/cm³ (20 °C)
EINECS:205-568-5
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤1.5 % (K.F.)
- Solubility:(25 % in water at 25°C) Clear colorless to light yellow
- UV:(0.005 M in water) 1 cm cell
- Titration after Ion exchange:≥98.5 % (On dry substance)
Specification Test Results
Frequently Bought Together