Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanedithiol ≥98%

1,8-Octanedithiol ≥98%

1,8-Octanedithiol ≥98%
Certificates
1,8-Octanedithiol ≥98%
1,8-Octanedithiol ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 1191-62-4 Molecular Formula: HS(CH2)8SH Formula Weight: 178.36 Flash Point: 113°(235°F) Refractive Index: 1.5050 Odour: Unpleasant Boiling Point: 269-270° Beilstein Reference: 1735431 MDL No.: MFCD00003574

Synonyms:  1,8-Dimercaptooctane
  • CAS Number:1191-62-4
  • Formula:C₈H₁₈S₂
  • MDL Number:MFCD00003574
  • Molecular Weight:178.36 g/mol
  • Boiling Pt:269…270 °C
  • Density:0.960
  • Flash Pt:113 °C (235 °F)
  • Storage Temperature:Ambient
  • EINECS:214-738-8
  • UN:3334
  • ADR:9,
  •  :Density: 0.9814 g/cm³ (0 °C)
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.