Order Entry
Poland
Orders LinkContactUsLinkComponent
 

1,8-Octanedithiol 99%

Irritant
1,8-Octanedithiol 99%
Certificates
1,8-Octanedithiol 99%

2 Options Available Below

Product Details & Documents
Synonyms:  1,8-Dimercaptooctane
  • CAS Number:1191-62-4
  • Formula:C₈H₁₈S₂
  • MDL Number:MFCD00003574
  • Molecular Weight:178.36 g/mol
  • Boiling Pt:269…270 °C (760 mmHg)
  • Density:0.96 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:214-738-8
  • UN:3334
  • ADR:9,III
  •  :Density: 0.9814 g/cm³ (0 °C)

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5030 to 1.5050 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 220 zł
    220 zł
    1 * 1 g
       220 zł

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 490 zł
    490 zł
    1 * 5 g
       490 zł