


Ethanethiol 99+%
:75-08-1
:
:C₂H₆S
:MFCD00004887
:12,03771
:35 °C (760 mmHg)
:–148 °C
:0.83 g/cm³
:–45 °C
:Ambient
:2363
:3,I
:62.14 g/mol
:200-837-3
:GHS07
:GHS09
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4296 to 1.4316 (20°C, 589 nm)
- GC:≥99.0 %
- Cloud point:≤20.0°C
- Calcium (Ca):≤30 ppm
- Iron (Fe):≤10 ppm
- Sodium (Na):≤200 ppm
- Insoluble matter:≤0.005 %
Frequently Bought Together