Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanediol ≥98%

1,8-Octanediol ≥98%

1,8-Octanediol ≥98%
Certificates
1,8-Octanediol ≥98%
1,8-Octanediol ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 629-41-4 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 148°(298°F) Melting Point: 58-61° Boiling Point: 171-173°/20mm Beilstein Reference: 1633499 MDL No.: MFCD00002989

Synonyms:  1,8-Dihydroxy octane
  • CAS Number:629-41-4
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00002989
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:171…173 °C (20 mmHg)
  • Melting Pt:58…61 °C
  • Density:0.941 g/cm³ (20 °C)
  • Flash Pt:148 °C (298 °F)
  • Storage Temperature:Ambient
  • EINECS:211-090-8
Product Family Options

Product Information