Order Entry
Poland
ContactUsLinkComponent
 
Methanol, SupraSolv® for gas chromatography, Supelco®
 :  Merck
    
 
undefined
Methanol, SupraSolv® for gas chromatography, Supelco®
 :  Merck
    
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -98 °C
  • Molar Mass:
    32.04 g/mol
  • Boiling Point:
    64.5 °C (1013 hPa)
  • Vapor Pressure:
    128 hPa (20 °C)
  • Flash Point:
    10 °C
  • Refractive Index:
    1.33
  • Explosion Limit:
    5.5 - 44 %(V)
  • Density:
    0.792 g/cm3 (20 °C)
  • pH:
    (H2O) no data available
  • Ignition Point:
    455 °C DIN 51794
  • Evaporation number:
    6.3
  • Water absorption:
    1000 g/kg
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

 

Frequently Bought Together