

1-Undecanol for synthesis, Sigma-Aldrich®
Catalog # 8.08468.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:13 - 15 °C
- Molar Mass:172.31 g/mol
- Boiling Point:129 - 131 °C (16 hPa)
- Flash Point:113 °C
- Refractive Index:1.44 (20 °C, 589 nm)
- Density:0.83 g/cm3 (20 °C)
- pH:7 (H2O, 20 °C)
- Synonyms:Undecyl alcohol
- Hill Formula:C11H24O
- Chemical Formula:CH3(CH2)10OH
Specification Test Results
About this item
:
:172.31 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:13…15 °C
Density:0.83 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-970-5
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
CAS Number:112-42-5
Formula:C₁₁H₂₄O
MDL Number:MFCD00004751