
1-Dodecanol, Supelco®
Catalog # 44095-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:186.34 g/mol
Merck Index:13,03439
Boiling Pt:260 °C (1013 hPa)
Melting Pt:22…24 °C
Density:0.81976 g/cm³ (20 °C)
Flash Pt:119 °C
Storage Temperature:Ambient
EINECS:203-982-0
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
:GHS09
CAS Number:112-53-8
Formula:H₃C(CH₂)₁₀CH₂OH
MDL Number:MFCD00004753