

1-Dodecanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-53-8
:
Formula:H₃C(CH₂)₁₀CH₂OH
MDL Number:MFCD00004753
:186.34 g/mol
Merck Index:13,03439
Boiling Pt:258…265 °C (760 mmHg)
Melting Pt:22…27 °C
Density:0.835 g/cm³
Flash Pt:119 °C
Storage Temperature:Ambient
EINECS:203-982-0
UN:3077
ADR:9,III
:GHS07
:GHS09
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
Specifications
Specification Test Results
- Appearance:Colorless to white Liquid or crystalline solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- HPLC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together